missing translation for 'onlineSavingsMsg'
Learn More

beta-Bromo-4-nitrophenetole 97.0+%, TCI America™

Catalog No. p-7122837
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32541G 1 g
B32545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32541G Supplier TCI America Supplier No. B32541G
Only null left

Chemical Identifiers

CAS 13288-06-7
Molecular Formula C8H8BrNO3
Molecular Weight (g/mol) 246.06
MDL Number MFCD00031363
InChI Key YQWCBDNNEZHPMA-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-nitrobenzene, 2-bromoethyl 4-nitrophenyl ether, beta-bromo-4-nitrophenetole, 2-4-nitrophenoxy ethyl bromide, 4-2-bromoethoxy-1-nitrobenzene, 1-2-bromoethyloxy-4-nitro-benzene, benzene, 1-2-bromoethoxy-4-nitro, 4-2-bromoethoxy nitrobenzene, acmc-1azdi
PubChem CID 235987
IUPAC Name 1-(2-bromoethoxy)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(OCCBr)C=C1

Specifications

Melting Point 66°C
Color Yellow
Boiling Point 170°C
Quantity 1 g
Formula Weight 246.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Bromo-4-nitrophenetole