missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Besifloxacin hydrochloride

Catalog No. AC467501000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467501000 100 mg
AC467505000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467501000 Supplier Thermo Scientific Chemicals Supplier No. 467501000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 405165-61-9
Molecular Formula C19H21ClFN3O3·HCl

Specifications

Quantity 100 mg
Formula Weight 430.31
InChI Key PMQBICKXAAKXAY-UHFFFAOYNA-N
SMILES Cl.NC1CCCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1Cl)C(O)=O
IUPAC Name 7-(3-aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Molecular Weight (g/mol) 430.30
Chemical Name or Material Besifloxacin hydrochloride

RUO – Research Use Only