missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BES, 0.5M buffer soln., pH 6.0, Thermo Scientific™
$145.73 - $145.73
Chemical Identifiers
| CAS | 66992-27-6 |
|---|---|
| Molecular Formula | C6H14NNaO5S |
| Molecular Weight (g/mol) | 235.23 |
| MDL Number | MFCD00067425 |
| InChI Key | CFQLQLSIZOWFNV-UHFFFAOYSA-M |
| PubChem CID | 23671640 |
| IUPAC Name | sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate |
| SMILES | C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60058AK
|
Thermo Scientific Chemicals
J60058AK |
250 mL |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 66992-27-6 | |
| 235.23 | |
| CFQLQLSIZOWFNV-UHFFFAOYSA-M | |
| sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate |
| C6H14NNaO5S | |
| MFCD00067425 | |
| 23671640 | |
| C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+] |
Specifications
| 66992-27-6 | |
| 0.5 M | |
| Liquid | |
| C6H14NNaO5S | |
| CFQLQLSIZOWFNV-UHFFFAOYSA-M | |
| sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate | |
| 23671640 |
| Colorless | |
| 6.0 | |
| 250 mL | |
| MFCD00067425 | |
| C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+] | |
| 235.23 | |
| BES buffer soln. |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold