missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium Bromide 98.0+%, TCI America™

$69.31 - $69.31

Chemical Identifiers

CAS 1449-46-3
Molecular Formula C25H22BrP
Molecular Weight (g/mol) 433.33
MDL Number MFCD00031707
InChI Key WTEPWWCRWNCUNA-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium bromide, bromo benzyl triphenylphosphorane, benzyltriphenylphosphanium bromide, phenylmethyl triphenylphosphonium bromide, benzyl triphenylphosphonium bromide, benzyl triphenyl phosphonium bromide, phosphonium, triphenyl phenylmethyl-, bromide, acmc-1ch0e, ksc491k3n
PubChem CID 2734970
IUPAC Name benzyltriphenylphosphanium bromide
SMILES [Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B202525G
View Documents
TCI America
B202525G
25 g
Each for $69.31
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1449-46-3
433.33
WTEPWWCRWNCUNA-UHFFFAOYSA-M
2734970
[Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C25H22BrP
MFCD00031707
benzyltriphenylphosphonium bromide, bromo benzyl triphenylphosphorane, benzyltriphenylphosphanium bromide, phenylmethyl triphenylphosphonium bromide, benzyl triphenylphosphonium bromide, benzyl triphenyl phosphonium bromide, phosphonium, triphenyl phenylmethyl-, bromide, acmc-1ch0e, ksc491k3n
benzyltriphenylphosphanium bromide

Specifications

1449-46-3
C25H22BrP
25 g
WTEPWWCRWNCUNA-UHFFFAOYSA-M
benzyltriphenylphosphanium bromide
2734970
≥98.0% (HPLC,T)
Benzyltriphenylphosphonium Bromide
White
MFCD00031707
benzyltriphenylphosphonium bromide, bromo benzyl triphenylphosphorane, benzyltriphenylphosphanium bromide, phenylmethyl triphenylphosphonium bromide, benzyl triphenylphosphonium bromide, benzyl triphenyl phosphonium bromide, phosphonium, triphenyl phenylmethyl-, bromide, acmc-1ch0e, ksc491k3n
[Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
433.33
433.33
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2608

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'