accessibility menu, dialog, popup

UserName

BrP

CAS RN 1449-46-3

IUPAC Name: benzyltriphenylphosphanium bromide
Synonyms: benzyltriphenylphosphanium bromide
Molecular Weight (g/mol): 433.33
Molecular Formula: C25H22BrP
InChi Key: WTEPWWCRWNCUNA-UHFFFAOYSA-M
SMILES: [Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)