missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltrimethylammonium Bromide 98.0+%, TCI America™
$61.51 - $61.51
Chemical Identifiers
| CAS | 5350-41-4 |
|---|---|
| Molecular Formula | C10H16BrN |
| Molecular Weight (g/mol) | 230.149 |
| MDL Number | MFCD00011780 |
| InChI Key | UUZYBYIOAZTMGC-UHFFFAOYSA-M |
| Synonym | benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm |
| PubChem CID | 21449 |
| IUPAC Name | benzyl(trimethyl)azanium;bromide |
| SMILES | C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
Chemical Identifiers
| 5350-41-4 | |
| 230.149 | |
| UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
| 21449 | |
| C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
| C10H16BrN | |
| MFCD00011780 | |
| benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
| benzyl(trimethyl)azanium;bromide |
Specifications
| 5350-41-4 | |
| White-Yellow | |
| MFCD00011780 | |
| benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
| C[N+](C)(C)CC1=CC=CC=C1.[Br-] | |
| 230.149 | |
| 230.15 | |
| Crystalline Powder |
| 229°C | |
| C10H16BrN | |
| 25 g | |
| UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
| benzyl(trimethyl)azanium;bromide | |
| 21449 | |
| ≥98.0% (HPLC,T) | |
| Benzyltrimethylammonium Bromide |
Safety and Handling
EINECSNumber : (3)-2694
RTECSNumber : BO8392000
TSCA : No