missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium Bromide 98.0+%, TCI America™

Supplier:  TCI America B0443500G

 View more versions of this product

Catalog No. B0443500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyltriethylammonium Bromide
195°C
C13H22BrN
500 g
CHQVQXZFZHACQQ-UHFFFAOYSA-M
benzyltriethylazanium bromide
165294
≥98.0% (T)
5197-95-5
White
MFCD00011822
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
272.23
272.23
Crystalline Powder

Chemical Identifiers

5197-95-5
272.23
CHQVQXZFZHACQQ-UHFFFAOYSA-M
165294
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
C13H22BrN
MFCD00011822
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
benzyltriethylazanium bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2694

TSCA : No

SDS
Documents

Documents

Promotions

Promotions