accessibility menu, dialog, popup

UserName

BrCH3NH3CCH3

CAS RN 5197-95-5

IUPAC Name: benzyltriethylazanium bromide
Synonyms: benzyltriethylazanium bromide
Molecular Weight (g/mol): 272.23
Molecular Formula: C13H22BrN
InChi Key: CHQVQXZFZHACQQ-UHFFFAOYSA-M
SMILES: [Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)