missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium Bromide 98.0+%, TCI America™

Catalog No. B044325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044325G 25 g
B0443500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B044325G Supplier TCI America Supplier No. B044325G

Chemical Identifiers

CAS 5197-95-5
Molecular Formula C13H22BrN
Molecular Weight (g/mol) 272.23
MDL Number MFCD00011822
InChI Key CHQVQXZFZHACQQ-UHFFFAOYSA-M
Synonym benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
PubChem CID 165294
IUPAC Name benzyltriethylazanium bromide
SMILES [Br-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point 195°C
Color White
Quantity 25 g
Formula Weight 272.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium Bromide
EINECSNumber (3)-2694
TSCA No