missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium bromide, 98+%

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol

Supplier:  Thermo Scientific Chemicals A1429136

 View more versions of this product

Catalog No. AAA1429136


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzyltriethylammonium bromide
∼200°C (decomposition)
MFCD00011822
3739113
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
272.23
272.24
5197-95-5
C13H22BrN
500 g
Hygroscopic
CHQVQXZFZHACQQ-UHFFFAOYSA-M
benzyl(triethyl)azanium;bromide
165294
≥98%

Chemical Identifiers

5197-95-5
272.23
CHQVQXZFZHACQQ-UHFFFAOYSA-M
165294
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
C13H22BrN
MFCD00011822
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
benzyl(triethyl)azanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-986-1

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only