missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium bromide, 98+%

Catalog No. AAA1429114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429114 25 g
AAA1429122 100 g
AAA1429136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429114 Supplier Thermo Scientific Chemicals Supplier No. A1429114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5197-95-5
Molecular Formula C13H22BrN
Molecular Weight (g/mol) 272.23
MDL Number MFCD00011822
InChI Key CHQVQXZFZHACQQ-UHFFFAOYSA-M
Synonym benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
PubChem CID 165294
IUPAC Name benzyl(triethyl)azanium;bromide
SMILES [Br-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point ∼200°C (decomposition)
Quantity 25 g
Beilstein 3739113
Sensitivity Hygroscopic
Formula Weight 272.24
Percent Purity ≥98%
Chemical Name or Material Benzyltriethylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-986-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only