missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium Borohydride 90.0+%, TCI America™

Catalog No. p-7119970
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31285G 5 g
B312825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31285G Supplier TCI America Supplier No. B31285G

Chemical Identifiers

CAS 85874-45-9
Molecular Formula C13H22BN
Molecular Weight (g/mol) 203.136
MDL Number MFCD00191785
InChI Key HQGWCEJYXHFBSA-UHFFFAOYSA-N
Synonym benzyltriethylammonium borohydride, benzyltriethylazanium ? 1-boranuidylidene, n,n,n-triethyl-benzenemethanaminium tetrahydroborate
PubChem CID 10899792
IUPAC Name benzyl(triethyl)azanium;boron(1-)
SMILES [B-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 5 g
UN Number 3131
Formula Weight 207.17
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium Borohydride