missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde, 95%, stabilized

CAS: 60656-87-3 | C9H10O2 | 150.18 g/mol

$100.89 - $100.89

Chemical Identifiers

CAS 60656-87-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00191779
InChI Key NFNOAHXEQXMCGT-UHFFFAOYSA-N
Synonym 2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
PubChem CID 108989
SMILES O=CCOCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC376070010
SDS View Documents
Thermo Scientific Chemicals
376070010
1 g
Each for $100.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60656-87-3
150.18
NFNOAHXEQXMCGT-UHFFFAOYSA-N
108989
C9H10O2
MFCD00191779
2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
O=CCOCC1=CC=CC=C1

Specifications

60656-87-3
118°C to 120°C (13.0 mmHg)
94% min. (GC)
C9H10O2
MFCD00191779
06,III,1476
2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
NFNOAHXEQXMCGT-UHFFFAOYSA-N
2-phenylmethoxyacetaldehyde
108989
95%
1.0690g/mL
113°C
Glass Bottle
C6H5CH2OCH2CHO
1 g
1.069
Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
O=CCOCC1=CC=CC=C1
150.18
150.18
Benzyloxyacetaldehyde
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 262-349-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only