accessibility menu, dialog, popup

UserName

OO

CAS RN 60656-87-3

IUPAC Name: 2-(benzyloxy)acetaldehyde
Synonyms: 2-(benzyloxy)acetaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: NFNOAHXEQXMCGT-UHFFFAOYSA-N
SMILES: O=CCOCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)