missing translation for 'onlineSavingsMsg'
Learn More

(Benzyloxy)acetaldehyde (stabilized with Catechol) 95.0+%, TCI America™

Catalog No. p-7119678
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24811G 1 g
B24815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24811G Supplier TCI America Supplier No. B24811G
Only null left

Chemical Identifiers

CAS 60656-87-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00191779
InChI Key NFNOAHXEQXMCGT-UHFFFAOYSA-N
Synonym 2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
PubChem CID 108989
IUPAC Name 2-(benzyloxy)acetaldehyde
SMILES O=CCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Benzyloxy)acetaldehyde (stabilized with Catechol)
TSCA No
Recommended Storage Refrigerator