Learn More
Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals
$185.88 - $185.88
Chemical Identifiers
| CAS | 6921-34-2 |
|---|---|
| Molecular Formula | C7H7ClMg |
| Molecular Weight (g/mol) | 150.89 |
| MDL Number | MFCD00000469 |
| InChI Key | NRAFPLGJPPJUNB-UHFFFAOYSA-M |
| Synonym | benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium |
| PubChem CID | 2733352 |
| IUPAC Name | benzyl(chloro)magnesium |
| SMILES | Cl[Mg]CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389551000
|
Thermo Scientific Chemicals
389551000 |
100 mL | AcroSeal™ Glass bottle |
Each for $185.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6921-34-2 | |
| 150.89 | |
| NRAFPLGJPPJUNB-UHFFFAOYSA-M | |
| 2733352 | |
| Cl[Mg]CC1=CC=CC=C1 |
| C7H7ClMg | |
| MFCD00000469 | |
| benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium | |
| benzyl(chloro)magnesium |
Specifications
| Brown to Black | |
| 20.51,75.83 | |
| C7H7ClMg | |
| MFCD00000469 | |
| 1.0300g/mL | |
| Solubility in water: reacts. | |
| Cl[Mg]CC1=CC=CC=C1 | |
| 150.89 | |
| 150.89 | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Iridescent Powder or Crystals |
| 6921-34-2 , 109-99-9 | |
| 24.17,79.49 | |
| (C6H5)CH2MgCl | |
| 1.40 to 1.65M | |
| benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium | |
| NRAFPLGJPPJUNB-UHFFFAOYSA-M | |
| benzyl(chloro)magnesium | |
| 2733352 | |
| 19 to 21% (as BMC) (Total base) | |
| 1.03 | |
| −17°C | |
| Benzylmagnesium chloride, 1.5M solution in THF |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
RUO – Research Use Only