missing translation for 'onlineSavingsMsg'
Learn More

Benzyldodecyldimethylammonium Bromide 97.0+%, TCI America™

Catalog No. p-7123141
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37485G 5 g
B374825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37485G Supplier TCI America Supplier No. B37485G
Only null left

Chemical Identifiers

CAS 7281-04-1
Molecular Formula C21H38BrN
Molecular Weight (g/mol) 384.45
MDL Number MFCD00011768
InChI Key KHSLHYAUZSPBIU-UHFFFAOYSA-M
PubChem CID 23705
IUPAC Name benzyl-dodecyl-dimethylazanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point 54°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 384.45
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldodecyldimethylammonium Bromide
RTECSNumber BO7875000
TSCA Yes