missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyldodecyldimethylammonium Bromide 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 2811 |
| Formula Weight | 384.45 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzyldodecyldimethylammonium Bromide |
Safety and Handling
| RTECSNumber | BO7875000 |
| TSCA | Yes |