missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyldimethyltetradecylammonium Chloride Hydrate 98.0+%, TCI America™
Supplier: TCI America A020825G
Specifications
| Benzyldimethyltetradecylammonium Chloride Hydrate | |
| White | |
| MFCD00011771 | |
| 2923 | |
| OCBHHZMJRVXXQK-UHFFFAOYSA-M | |
| benzyldimethyltetradecylazanium chloride | |
| 8755 | |
| ≥98.0% (T) |
| 139-08-2 | |
| C23H42ClN | |
| 25 g | |
| miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg | |
| [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 | |
| 368.05 | |
| 368.05 | |
| Crystalline Powder |
Chemical Identifiers
| 139-08-2 | |
| 368.05 | |
| OCBHHZMJRVXXQK-UHFFFAOYSA-M | |
| 8755 | |
| [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
| C23H42ClN | |
| MFCD00011771 | |
| miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg | |
| benzyldimethyltetradecylazanium chloride |
Safety and Handling
EINECSNumber : (3)-2694
RTECSNumber : BO7150000
TSCA : Yes