accessibility menu, dialog, popup

UserName

ClH3CNCH3H3C

CAS RN 139-08-2

IUPAC Name: benzyldimethyltetradecylazanium chloride
Synonyms: miristalkonium chloride|myristalkonium chloride|tetradecyl dimethyl benzyl ammonium chloride|alkyl (95%C14, 3%C12, 2%C16) dimethyl benzyl ammonium chloride|alkyl (60%C14, 25%C12, 15%C16) dimethylbenzyl ammonium chloride|alkyl* dimethyl benzyl ammonium chloride *(93%C14, 4%C12, 3%C16)|alkyl* dimethyl benzyl ammonium chloride *(40%C12, 40%C14, 20%C16)|alkyl* dimethyl benzyl ammonium chloride *(65%C12, 25%C14, 10%C16)|alkyl* dimethyl benzyl ammonium chloride *(60% C14, 30% C16, 10% C12)|alkyl* dimethyl benzyl ammonium chloride *(67%C12, 25%C14, 7%C16, 1%C18)
Molecular Weight (g/mol): 368.05
Molecular Formula: C23H42ClN
InChi Key: OCBHHZMJRVXXQK-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)