missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyltetradecylammonium Chloride Hydrate 98.0+%, TCI America™

Catalog No. p-7116767
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A020825G 25 g
A0208100G 100 g
A0208500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A020825G Supplier TCI America Supplier No. A020825G
Only null left

Chemical Identifiers

CAS 139-08-2
Molecular Formula C23H42ClN
Molecular Weight (g/mol) 368.05
MDL Number MFCD00011771
InChI Key OCBHHZMJRVXXQK-UHFFFAOYSA-M
Synonym miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
PubChem CID 8755
IUPAC Name benzyldimethyltetradecylazanium chloride
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Color White
Quantity 25 g
UN Number 2923
Formula Weight 368.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethyltetradecylammonium Chloride Hydrate
EINECSNumber (3)-2694
RTECSNumber BO7150000
TSCA Yes