Learn More
Benzyldimethyl-n-dodecylammonium chloride, 98%
CAS: 139-07-1 | C21H38ClN | 339.99 g/mol
$113.15 - $847.34
Chemical Identifiers
| CAS | 139-07-1 |
|---|---|
| Molecular Formula | C21H38ClN |
| Molecular Weight (g/mol) | 339.99 |
| MDL Number | MFCD00137276 |
| InChI Key | JBIROUFYLSSYDX-UHFFFAOYSA-M |
| Synonym | benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl |
| PubChem CID | 8753 |
| IUPAC Name | benzyl-dodecyl-dimethylazanium;chloride |
| SMILES | [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6019606
|
Thermo Scientific Chemicals
H6019606 |
5 g |
Each for $113.15
|
|
|||||
|
AAH6019618
|
Thermo Scientific Chemicals
H6019618 |
50 g |
Each for $847.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 139-07-1 | |
| 339.99 | |
| JBIROUFYLSSYDX-UHFFFAOYSA-M | |
| 8753 | |
| [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
| C21H38ClN | |
| MFCD00137276 | |
| benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl | |
| benzyl-dodecyl-dimethylazanium;chloride |
Specifications
| 139-07-1 | |
| C21H38ClN | |
| 5 g | |
| 3921689 | |
| JBIROUFYLSSYDX-UHFFFAOYSA-M | |
| benzyl-dodecyl-dimethylazanium;chloride | |
| 8753 | |
| 98% |
| 60°C | |
| MFCD00137276 | |
| UN3261 | |
| benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl | |
| [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 | |
| 339.99 | |
| 339.99 | |
| Benzyldimethyl-n-dodecylammonium chloride |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 205-351-5
RTECSNumber : BO6821500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only