missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyl-n-dodecylammonium chloride, 98%

CAS: 139-07-1 | C21H38ClN | 339.99 g/mol

Supplier:  Thermo Scientific Chemicals H6019618

 View more versions of this product

Catalog No. AAH6019618


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzyldimethyl-n-dodecylammonium chloride
60°C
MFCD00137276
UN3261
benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
339.99
339.99
139-07-1
C21H38ClN
50 g
3921689
JBIROUFYLSSYDX-UHFFFAOYSA-M
benzyl-dodecyl-dimethylazanium;chloride
8753
98%

Chemical Identifiers

139-07-1
339.99
JBIROUFYLSSYDX-UHFFFAOYSA-M
8753
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
C21H38ClN
MFCD00137276
benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl
benzyl-dodecyl-dimethylazanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 205-351-5

RTECSNumber : BO6821500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only