missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B040725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B040725G 25 g
B0407100G 100 g
B0407500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B040725G Supplier TCI America Supplier No. B040725G

Chemical Identifiers

CAS 3287-99-8
Molecular Formula C7H10ClN
Molecular Weight (g/mol) 143.614
MDL Number MFCD00012852
InChI Key XKXHCNPAFAXVRZ-UHFFFAOYSA-N
Synonym benzylamine hydrochloride, phenylmethanamine hydrochloride, benzenemethanamine, hydrochloride, benzylammonium chloride, benzylamine, hydrochloride, usaf el-82, benzenemethanamine, hydrochloride 1:1, benzylamine hcl, benzylaminehydrochloride, benzyl amine hcl salt
PubChem CID 2724127
IUPAC Name phenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CN.Cl

Specifications

Melting Point 253°C
Color White
Quantity 25 g
Formula Weight 143.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylamine Hydrochloride
EINECSNumber (3)-0367
RTECSNumber DP5425000
TSCA Yes