accessibility menu, dialog, popup

UserName

CAS RN 3287-99-8

IUPAC Name: phenylmethanaminium chloride
Synonyms: benzylaminium chloride
Molecular Weight (g/mol): 143.61
Molecular Formula: C7H10ClN
InChi Key: XKXHCNPAFAXVRZ-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)