missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylamine Hydrobromide 98.0+%, TCI America™
$42.36 - $167.49
Chemical Identifiers
| CAS | 37488-40-7 |
|---|---|
| Molecular Formula | C7H10BrN |
| Molecular Weight (g/mol) | 188.068 |
| MDL Number | MFCD00035434 |
| InChI Key | QJFMCHRSDOLMHA-UHFFFAOYSA-N |
| Synonym | Benzylammonium Bromide |
| PubChem CID | 12998568 |
| IUPAC Name | phenylmethanamine;hydrobromide |
| SMILES | C1=CC=C(C=C1)CN.Br |
Chemical Identifiers
| 37488-40-7 | |
| 188.068 | |
| QJFMCHRSDOLMHA-UHFFFAOYSA-N | |
| 12998568 | |
| C1=CC=C(C=C1)CN.Br |
| C7H10BrN | |
| MFCD00035434 | |
| Benzylammonium Bromide | |
| phenylmethanamine;hydrobromide |
Specifications
| 37488-40-7 | |
| White | |
| MFCD00035434 | |
| Benzylammonium Bromide | |
| C1=CC=C(C=C1)CN.Br | |
| 188.068 | |
| 188.07 | |
| Crystalline Powder |
| 204°C | |
| C7H10BrN | |
| 1 g | |
| QJFMCHRSDOLMHA-UHFFFAOYSA-N | |
| phenylmethanamine;hydrobromide | |
| 12998568 | |
| ≥98.0% (HPLC,T) | |
| Benzylamine Hydrobromide |
Safety and Handling
TSCA : No