accessibility menu, dialog, popup

UserName

CAS RN 37488-40-7

H2NHBr

CAS RN 37488-40-7

IUPAC Name: 1-phenylmethanamine hydrobromide
Synonyms: benzylamine hydrobromide
Molecular Weight (g/mol): 188.07
Molecular Formula: C7H10BrN
InChi Key: QJFMCHRSDOLMHA-UHFFFAOYSA-N
SMILES: Br.NCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results