missing translation for 'onlineSavingsMsg'
Learn More

Benzylamine Hydrobromide 98.0+%, TCI America™

Catalog No. B51851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51851G 1 g
B51855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51851G Supplier TCI America Supplier No. B51851G
Only null left

Chemical Identifiers

CAS 37488-40-7
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.068
MDL Number MFCD00035434
InChI Key QJFMCHRSDOLMHA-UHFFFAOYSA-N
Synonym Benzylammonium Bromide
PubChem CID 12998568
IUPAC Name phenylmethanamine;hydrobromide
SMILES C1=CC=C(C=C1)CN.Br

Specifications

Melting Point 204°C
Color White
Quantity 1 g
Formula Weight 188.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylamine Hydrobromide