missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Salicylate 99.0+%, TCI America™
Supplier: TCI America S000925G
Specifications
| Benzyl Salicylate | |
| Yellow | |
| C14H12O3 | |
| 25 g | |
| ZCTQGTTXIYCGGC-UHFFFAOYSA-N | |
| benzyl 2-hydroxybenzoate | |
| 8363 | |
| ≥99.0% (GC) |
| 118-58-1 | |
| 194°C | |
| MFCD00020034 | |
| benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester | |
| C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O | |
| 228.247 | |
| 228.25 | |
| Liquid |
Chemical Identifiers
| 118-58-1 | |
| 228.247 | |
| ZCTQGTTXIYCGGC-UHFFFAOYSA-N | |
| 8363 | |
| C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O |
| C14H12O3 | |
| MFCD00020034 | |
| benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester | |
| benzyl 2-hydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-1570&(3)-3044
RTECSNumber : VO1750000
TSCA : Yes