missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Salicylate 99.0+%, TCI America™

Catalog No. p-7135760
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000925G 25 g
S0009500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000925G Supplier TCI America Supplier No. S000925G
Only null left

Chemical Identifiers

CAS 118-58-1
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00020034
InChI Key ZCTQGTTXIYCGGC-UHFFFAOYSA-N
Synonym benzyl salicylate, benzyl o-hydroxybenzoate, salicylic acid, benzyl ester, benzoic acid, 2-hydroxy-, phenylmethyl ester, phenylmethyl 2-hydroxybenzoate, salicylic acid benzyl ester, salicyclic acid, benzyl ester, salicylsaeurebenzylester, unii-wao5mnk9tu, salicyclic acid benzyl ester
PubChem CID 8363
IUPAC Name benzyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2O

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 g
Formula Weight 228.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Salicylate
EINECSNumber (3)-1570&(3)-3044
RTECSNumber VO1750000
TSCA Yes