missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(+)-Glycidyl Ether 98.0+%, TCI America™

Catalog No. B22391G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22391G 1 g
B22395G 5 g
B223925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22391G Supplier TCI America Supplier No. B22391G

Chemical Identifiers

CAS 16495-13-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00054428
InChI Key QNYBOILAKBSWFG-SNVBAGLBSA-N
Synonym s-+-benzyl glycidyl ether, s-benzyloxymethyl-oxirane, benzyl s-+-glycidyl ether, s-2-benzyloxy methyl oxirane, 2s-2-benzyloxy methyl oxirane, s-o-benzylglycidol, s-benzyl glycidyl ether, +-benzyl glycidyl ether, s-benzyloxymethyl oxirane
PubChem CID 146296
IUPAC Name (2S)-2-[(benzyloxy)methyl]oxirane
SMILES C(OCC1=CC=CC=C1)[C@@H]1CO1

Specifications

Color Colorless
Boiling Point 130°C
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl (S)-(+)-Glycidyl Ether
RTECSNumber TX2860020
TSCA No