accessibility menu, dialog, popup

UserName

CAS RN 16495-13-9

IUPAC Name: (2S)-2-[(benzyloxy)methyl]oxirane
Synonyms: (2S)-2-[(benzyloxy)methyl]oxirane
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: QNYBOILAKBSWFG-SNVBAGLBSA-N
SMILES: C(OCC1=CC=CC=C1)[C@@H]1CO1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)