missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(+)-glycidyl ether, 98+%

Catalog No. p-7046347
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1403303 1 g
AAL1403306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1403303 Supplier Thermo Scientific Chemicals Supplier No. L1403303

CAS: 16495-13-9 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16495-13-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00054428
InChI Key QNYBOILAKBSWFG-SNVBAGLBSA-N
Synonym s-+-benzyl glycidyl ether, s-benzyloxymethyl-oxirane, benzyl s-+-glycidyl ether, s-2-benzyloxy methyl oxirane, 2s-2-benzyloxy methyl oxirane, s-o-benzylglycidol, s-benzyl glycidyl ether, +-benzyl glycidyl ether, s-benzyloxymethyl oxirane
PubChem CID 146296
SMILES C(OCC1=CC=CC=C1)[C@@H]1CO1

Specifications

Density 1.072
Boiling Point 130°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 1 g
Beilstein 5246495
Optical Rotation +5.1° (c=5 in Toluene)
IUPAC Name (2S)-2-[(benzyloxy)methyl]oxirane
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material Benzyl (S)-(+)-glycidyl ether
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber TX2860020
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only