missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Glycidyl Ether 97.0+%, TCI America™
$72.70 - $186.41
Chemical Identifiers
| CAS | 2930-05-4 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.20 |
| MDL Number | MFCD00068664 |
| InChI Key | QNYBOILAKBSWFG-UHFFFAOYNA-N |
| Synonym | 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane |
| PubChem CID | 94247 |
| IUPAC Name | 2-(phenylmethoxymethyl)oxirane |
| SMILES | C1C(O1)COCC2=CC=CC=C2 |
Chemical Identifiers
| 2930-05-4 | |
| 164.20 | |
| QNYBOILAKBSWFG-UHFFFAOYNA-N | |
| 94247 | |
| C1C(O1)COCC2=CC=CC=C2 |
| C10H12O2 | |
| MFCD00068664 | |
| 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane | |
| 2-(phenylmethoxymethyl)oxirane |
Specifications
| 2930-05-4 | |
| 75°C | |
| MFCD00068664 | |
| 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane | |
| C1C(O1)COCC2=CC=CC=C2 | |
| 164.20 | |
| 164.20 | |
| Liquid |
| Yellow | |
| C10H12O2 | |
| 5 g | |
| QNYBOILAKBSWFG-UHFFFAOYNA-N | |
| 2-(phenylmethoxymethyl)oxirane | |
| 94247 | |
| ≥97.0% (GC) | |
| Benzyl Glycidyl Ether |
Safety and Handling
RTECSNumber : TX2860000
TSCA : No
Recommended Storage : Refrigerator