CAS RN 2930-05-4
CAS RN 2930-05-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzyl Glycidyl Ether 97.0+%, TCI America™
CAS: 2930-05-4 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.20 MDL Number: MFCD00068664 InChI Key: QNYBOILAKBSWFG-UHFFFAOYNA-N Synonym: 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane PubChem CID: 94247 IUPAC Name: 2-(phenylmethoxymethyl)oxirane SMILES: C1C(O1)COCC2=CC=CC=C2