Learn More
Benzyl chloromethyl ether, tech. 70%
CAS: 3587-60-8 | C8H9ClO | 156.609 g/mol
$236.55 - $718.48
Chemical Identifiers
| CAS | 3587-60-8 |
|---|---|
| Molecular Formula | C8H9ClO |
| Molecular Weight (g/mol) | 156.609 |
| MDL Number | MFCD00000886 |
| InChI Key | LADPCMZCENPFGV-UHFFFAOYSA-N |
| Synonym | benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl |
| PubChem CID | 137983 |
| IUPAC Name | chloromethoxymethylbenzene |
| SMILES | C1=CC=C(C=C1)COCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH60207AD
|
Thermo Scientific Chemicals
H60207AD |
50 mL |
Each for $236.55
|
|
|||||
|
AAH60207AK
|
Thermo Scientific Chemicals
H60207AK |
250 mL |
Each for $718.48
|
|
|||||
Description
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.
Solubility
Soluble in water [2.7 g/L (25 C)]
Notes
Store at 2-8°C. Keep away from oxidizing agents.
Chemical Identifiers
| 3587-60-8 | |
| 156.609 | |
| LADPCMZCENPFGV-UHFFFAOYSA-N | |
| 137983 | |
| C1=CC=C(C=C1)COCCl |
| C8H9ClO | |
| MFCD00000886 | |
| benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl | |
| chloromethoxymethylbenzene |
Specifications
| 3587-60-8 | |
| 102°C | |
| 1.527 | |
| MFCD00000886 | |
| UN2810 | |
| benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl | |
| LADPCMZCENPFGV-UHFFFAOYSA-N | |
| chloromethoxymethylbenzene | |
| 137983 | |
| 70% | |
| Benzyl chloromethyl ether |
| 1.126 | |
| C8H9ClO | |
| C6H5CH2OCH2Cl | |
| 50 mL | |
| 1364034 | |
| Soluble in water [2.7g/L (25 C)] | |
| C1=CC=C(C=C1)COCCl | |
| 156.609 | |
| 156.61 | |
| Technical |
Safety and Handling
GHS H Statement
H331-H350-H373-H318-H302-H315-H335
Toxic if inhaled.
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H334-H335-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only