accessibility menu, dialog, popup

UserName

CAS RN 3587-60-8

IUPAC Name: [(chloromethoxy)methyl]benzene
Synonyms: [(chloromethoxy)methyl]benzene
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: LADPCMZCENPFGV-UHFFFAOYSA-N
SMILES: ClCOCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)