missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloromethyl ether, tech. 70%

CAS: 3587-60-8 | C8H9ClO | 156.609 g/mol

Supplier:  Thermo Scientific Chemicals H60207AK

 View more versions of this product

Catalog No. AAH60207AK


Only null left
Explore available promotions

Description

Description

Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

Solubility
Soluble in water [2.7 g/L (25 C)]

Notes
Store at 2-8°C. Keep away from oxidizing agents.
Specifications

Specifications

Benzyl chloromethyl ether
1.126
C8H9ClO
C6H5CH2OCH2Cl
250 mL
1364034
Soluble in water [2.7g/L (25 C)]
C1=CC=C(C=C1)COCCl
156.609
156.61
Technical
3587-60-8
102°C
1.527
MFCD00000886
UN2810
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
LADPCMZCENPFGV-UHFFFAOYSA-N
chloromethoxymethylbenzene
137983
70%

Chemical Identifiers

3587-60-8
156.609
LADPCMZCENPFGV-UHFFFAOYSA-N
137983
C1=CC=C(C=C1)COCCl
C8H9ClO
MFCD00000886
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
chloromethoxymethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H350-H373-H318-H302-H315-H335
Toxic if inhaled.
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H334-H335-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only