missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Carbamate 97.0+%, TCI America™
Supplier: TCI America C0590250G
| Quantity | 250 g |
|---|
Chemical Identifiers
| C8H9NO2 | |
| MFCD00007965 | |
| carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
| benzyl carbamate |
Specifications
| 87°C | |
| 621-84-1 | |
| C8H9NO2 | |
| carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
| NC(=O)OCC1=CC=CC=C1 | |
| 250 g | |
| 12136 | |
| ≥97.0% (N) |
| Benzyl Carbamate | |
| White | |
| MFCD00007965 | |
| PUJDIJCNWFYVJX-UHFFFAOYSA-N | |
| benzyl carbamate | |
| 151.17 | |
| 151.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No