accessibility menu, dialog, popup

UserName

H2NOO

CAS RN 621-84-1

IUPAC Name: benzyl carbamate
Synonyms: O-benzyl carbamate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: PUJDIJCNWFYVJX-UHFFFAOYSA-N
SMILES: NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)