missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Carbamate 97.0+%, TCI America™

Catalog No. p-7124516
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C059025G 25 g
C0590250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C059025G Supplier TCI America Supplier No. C059025G
Only null left

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 87°C
Color White
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Carbamate