missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Benzoate, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7157084
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600916 500 mL Amber Glass Bottle
18600917 4 L Amber Glass Bottle
18600918 20 L Amber Glass Bottle
18600919 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600916 Supplier Spectrum Chemical Mfg Cor Supplier No. BE159500MLGL
Only null left

BE159, 120-51-4, C14H12O2

Spectrum™ Chemical Benzyl Benzoate, USP is used as a antiparasitic insecticide against human scabies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 100%
Boiling Point 323°C
Packaging Amber Glass Bottle
Refractive Index 1.568-1.570
Quantity 500 mL
Percent Purity 99 to 100.5%
Grade USP
Physical Form Liquid