accessibility menu, dialog, popup

UserName

OO

CAS RN 120-51-4

IUPAC Name: benzyl benzoate
Synonyms: benzyl benzoate benzemul caswell no. 082 scabagen colebenz benzyl benzoate (product) scabanca peruscabina scabitox aventis brand of benzyl benzoate
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: SESFRYSPDFLNCH-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (4)
  • (2)
  • (5)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (9)
  • (4)