missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Benzoate 99.0+%, TCI America™

Catalog No. B006425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006425G 25 g
B0064500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B006425G Supplier TCI America Supplier No. B006425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
Synonym ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
PubChem CID 2345
ChEBI CHEBI:41237
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 324°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Benzoate
EINECSNumber (3)-1389
RTECSNumber DG4200000
TSCA Yes