missing translation for 'onlineSavingsMsg'
Learn More

Benzyl benzoate, 99+%

Catalog No. AAL032580E
Change view
Click to view available options
Quantity:
1000 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL032580E 2500 g
AAL032580B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL032580E Supplier Thermo Scientific Chemicals Supplier No. L032580E
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 120-51-4 | C14H12O2 | 212.25 g/mol

Benzyl benzoate is used as repellent for chiggers, ticks, and mosquitoes. It is an effective and inexpensive topical treatment for human scabies. It has vasodilating and spasmolytic effects and is present in many asthma and whooping cough drugs. It is also a dye carrier, solvent for cellulose derivatives, plasticizer, and fixative in the perfume industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with ethanol, alcohol, chloroform, ether, oils.

Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003075
InChI Key SESFRYSPDFLNCH-UHFFFAOYSA-N
Synonym ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
PubChem CID 2345
ChEBI CHEBI:41237
IUPAC Name benzyl benzoate
SMILES O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 18°C to 20°C
Density 1.112
Boiling Point 323°C to 324°C
Flash Point 147°C (296°F)
Refractive Index 1.568
Linear Formula C6H5CO2CH2C6H5
Quantity 2500 g
UN Number UN3082
Beilstein 2049280
Merck Index 14,1127
Solubility Information Miscible with ethanol,alcohol,chloroform,ether,oils.
Formula Weight 212.25
Percent Purity ≥99%
Chemical Name or Material Benzyl benzoate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 204-402-9
RTECSNumber DG4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only