Learn More
Benzyl benzoate, 99+%, Thermo Scientific Chemicals
Description
Benzyl benzoate is used as repellent for chiggers, ticks, and mosquitoes. It is an effective and inexpensive topical treatment for human scabies. It has vasodilating and spasmolytic effects and is present in many asthma and whooping cough drugs. It is also a dye carrier, solvent for cellulose derivatives, plasticizer, and fixative in the perfume industry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Miscible with ethanol, alcohol, chloroform, ether, oils.
Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
Specifications
Specifications
| Melting Point | 18°C to 20°C |
| Density | 1.112 |
| Boiling Point | 323°C to 324°C |
| Flash Point | 147°C (296°F) |
| Refractive Index | 1.568 |
| Linear Formula | C6H5CO2CH2C6H5 |
| Quantity | 1000 g |
| UN Number | UN3082 |
| Beilstein | 2049280 |
| Merck Index | 14,1127 |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 204-402-9 |
| RTECSNumber | DG4200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only