missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 205.3°C |
| Packaging | Amber Glass Bottle |
| Quantity | 20 L |
| Specific Gravity | 1.042 - 1.047 |
| Grade | Technical |
| Physical Form | Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.