missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Technical, Spectrum™ Chemical

Catalog No. 18609055
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609055 55 gal. Amber Glass Bottle
18609054 500 mL Amber Glass Bottle
18609058 4 L Amber Glass Bottle
18609057 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609055 Supplier Spectrum Chemical Mfg Cor Supplier No. B109255GLBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-51-6

Specifications

CAS Max % 1
Boiling Point 205.3°C
Packaging Amber Glass Bottle
Quantity 55 gal.
Specific Gravity 1.042 - 1.047
Grade Technical
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.