missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -15.4°C |
| Color | Colorless |
| Density | 1.045g/mL at 25°C |
| Boiling Point | 203°C to 205°C |
| Flash Point | 96°C (204.8°F) |
| Ignition Residue | ≤0.005% (as SO4) |
| Ignition Point | 817°F |
| Packaging | Glass Bottle |
| Refractive Index | n20/D 1.539(lit.) |
| Linear Formula | C6H5CH2OH |
| Show More |
Safety and Handling
| Hazard Statement | P261-P301 + P312 + P330-P305 + P351 + P338 |
| Precautionary Statement | H302 + H332-H319 |
| EINECSNumber | 202-859-9 |
| RTECSNumber | DN3150000 |