missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, puriss., 99 to 100.5% (GC), Honeywell Riedel-de Haën™

Catalog No. 60045515
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless steel can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045515 30 kg Stainless steel can
60045513 1 L Glass Bottle
60045514 2.5 L Glass Bottle
60045517 6 x 1 L Glass Bottle
60045516 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60045515 Supplier Honeywell Chemicals Supplier No. 2412230KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., NF, 99-100.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 203° to 205°C (lit.)
Melting Point -16°C to -13°C
Density 1.045g/mL at 25°C
Flash Point 96°C (204.8°F)
Ignition Residue ≤0.005% (as SO4)
Ignition Point 817°F
Packaging Stainless steel can
Linear Formula C6H5CH2OH
Quantity 30 kg
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Vapor Pressure 13.3mmHg (100°C), 3.75mmHg (77°C)
Formula Weight 108.14g/mol
Percent Purity 99 to 100.5% (GC)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, Meets analytical specification of Ph.Eur.BP, NF
Show More Show Less
Hazard Statement P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338
Precautionary Statement H302 + H332-H319
EINECSNumber 202-859-9
RTECSNumber DN3150000